Przejdź do
Z327085
BRAND® BLAUBRAND® volumetric flask, glass stopper, amber glass
volume 250 mL, accuracy: 0.15 mL
Synonim(y):
amber flask, analytical flask, calibration flask with stopper, measuring flask, volumetric flask
Wybierz wielkość
Zmień widok
Rozmiar/SKUDostępnośćCena netto
2 ea
Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności
378,00 zł
Informacje o tej pozycji
NACRES:
NB.43
UNSPSC Code:
41121804
eCl@ss:
32040501
Capacity:
250 mL
Material:
borosilicate glass 3.3 flask
Packaging:
pack of 2 ea
378,00 zł
Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności
​
Pomoc techniczna
Pozwól nam pomóc
material
borosilicate glass 3.3 flask
joint
joint ST/NS neck14/23
feature
class A DE-M, DIN EN ISO 1042. Calibrated to contain (TC, In).
packaging
pack of 2 ea
manufacturer/tradename
BRAND 37451
volume measuring range
250 mL, accuracy: 0.15 mL
capacity
250 mL
General description
Volumetric flasks according to DIN EN ISO 1042, Class A, conformity certified. Made from Boro 3.3, calibrated to contain (TC, IN). Supplied with a interchangeable amber glass stopper. Marks and inscriptions in high contrast white enamel.
All BLAUBRAND® volumetric flasks are supplied with a batch number and an accompanying batch certificate in the original packaging.
Other Notes
BR37451 (India)
Legal Information
BLAUBRAND is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG
Porównaj podobne pozycje
Wyświetl pełne porównanie
Pokaż różnice
1 of 1
Ta pozycja
Z327077Z326437Z326445
description
volume 250 mL, accuracy: 0.15 mL
description
volume 200 mL, accuracy: 0.15 mL
description
volume 50 mL, accuracy: 0.08 mL
description
volume 100 mL, accuracy: 0.1 mL
capacity
250 mL
capacity
200 mL
capacity
50 mL
capacity
100 mL
feature
class A DE-M, DIN EN ISO 1042. Calibrated to contain (TC, In).
feature
class A DE-M, DIN EN ISO 1042. Calibrated to contain (TC, In).
feature
class A DE-M, DIN EN ISO 1042. Calibrated to contain (TC, In).
feature
class A DE-M, DIN EN ISO 1042. Calibrated to contain (TC, In).
manufacturer/tradename
BRAND 37451
manufacturer/tradename
BRAND 37450
manufacturer/tradename
BRAND 37406
manufacturer/tradename
BRAND 37407
material
borosilicate glass 3.3 flask
material
borosilicate glass 3.3 flask
material
wide-neck , borosilicate glass 3.3 flask
material
borosilicate glass 3.3 flask
joint
joint ST/NS neck14/23
joint
joint ST/NS neck14/23
joint
joint ST/NS neck14/23
joint
joint ST/NS neck14/23
packaging
pack of 2 ea
packaging
pack of 2 ea
packaging
pack of 2 ea
packaging
pack of 2 ea
Still not finding the right product?
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)