Merck
Z326496
BRAND® BLAUBRAND® volumetric flask, PP stopper, amber glass
volume 1000 mL, accuracy: 0.4 mL
Synonim(y):
amber flask, analytical flask, calibration flask with stopper, measuring flask, volumetric flask
Informacje o tej pozycji
NACRES:
NB.43
UNSPSC Code:
41121804
eCl@ss:
32040501
Capacity:
1,000 mL
Material:
borosilicate glass 3.3 flask
Packaging:
pack of 1 ea
Pomoc techniczna
Pozwól nam pomóc
material
borosilicate glass 3.3 flask
joint
joint ST/NS neck24/29
description
calibrated to contain (TC, IN)
feature
class A DE-M, DIN EN ISO 1042. Calibrated to contain (TC, In).
packaging
pack of 1 ea
manufacturer/tradename
BRAND 37411
volume measuring range
1000 mL, accuracy: 0.4 mL
capacity
1,000 mL
General description
The BLAUBRAND® volumetric flask combines precision and reliability, featuring a square grip PP stopper for easy handling. Crafted from high-quality Boro 3.3 glass and calibrated to contain (TC, In), each flask is marked with high-contrast white enamel for clear readability. Supplied with a batch number and certificate, these flasks ensure quality and traceability, making them ideal for laboratory use.
Other Notes
BR37411 (India)
Legal Information
BLAUBRAND is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG
Still not finding the right product?
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)