Informacje o tej pozycji
NACRES:
NB.01
UNSPSC Code:
41121500
Capacity:
7 mL
Material:
AR-Glas®
Feature:
class AS
Packaging:
pack of 6 ea
Pomoc techniczna
Pozwól nam pomócmaterial
AR-Glas®
Quality Segment
description
total delivery Ex + 15 sec waiting time
feature
class AS
packaging
pack of 6 ea
manufacturer/tradename
BRAND 29709
capacity
7 mL
General description
Additional to DIN
All BLAUBRAND bulb pipettes are supplied with a batch number and an accompanying batch certificate in the original packaging.
According to DIN EN ISO 648.
According to DIN EN ISO 648.
Other Notes
BR29709 (India)
Legal Information
AR-Glas is a registered trademark of Schott AG
BLAUBRAND is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG
Still not finding the right product?
Explore all of our products under BRAND® BLAUBRAND® bulb pipette, calibrated to deliver (TD, EX)
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?