Merck
Z290610
Vacuum filtration assembly flasks
collection flask, capacity 1000 mL
Informacje o tej pozycji
NACRES:
NB.24
UNSPSC Code:
41121804
eCl@ss:
32040501
Capacity:
1000 mL
Material:
borosilicate glass , glass housing
Manufacturer/tradename:
Sigma-Aldrich
Packaging:
pack of 1, pkg of 1 ea
Pomoc techniczna
Pozwól nam pomóc
material
borosilicate glass , glass housing
sterility
non-sterile
packaging
pack of 1, pkg of 1 ea
manufacturer/tradename
Sigma-Aldrich
capacity
1000 mL
fitting
connector for 40/35 joint
General description
Flask does not have side arm.
Still not finding the right product?
Explore all of our products under Vacuum filtration assembly flasks
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)