Wybierz wielkość
| Rozmiar/SKU | Dostępność | Cena netto |
|---|---|---|
6 ea | Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności | 3630,00 zł 2904,00 zł |
Informacje o tej pozycji
2904,00 zł
Cena katalogowa3630,00 złZaoszczędź 20%material
polystyrene
sterility
sterile
feature
vented
packaging
case of 6 ea
manufacturer/tradename
Nunc 140250
surface area
1264 cm2
binding type
Nunclon Delta surface
General description
Features and Benefits
- "Out of the box" system with no accessories required.
- Large, vented screw closure for easy handling
- Versatile port design allows for pouring and aseptic filling.
- Fill and empty five times faster compared to T175 flasks.
- Enhances productivity by enabling faster operations.
- Fast start-up with a simple pour of media and cells into the large opening.
- Versatile port facilitates pouring and aseptic filling techniques.
Legal Information
1 of 1
Ta pozycja | |||
|---|---|---|---|
| description 2 layer, polystyrene, surface area 1264 cm2, sterile, vented, case of 6 ea | description 10 layer, polystyrene, surface area 6320 cm2, sterile, vented, case of 6 ea | description 4 layer, polystyrene, surface area 2528 cm2, sterile, vented, case of 4 ea | description 10 layer, polystyrene, surface area 6320 cm2, sterile, vented, case of 2 ea |
| feature vented | feature vented | feature vented | feature vented |
| material polystyrene | material polystyrene | material polystyrene | material polystyrene |
| sterility sterile | sterility sterile | sterility sterile | sterility sterile |
| binding type Nunclon Delta surface | binding type Nunclon Delta surface | binding type Nunclon Delta surface | binding type Nunclon Delta surface |
| surface area 1264 cm2 | surface area 6320 cm2 | surface area 2528 cm2 | surface area 6320 cm2 |
| packaging case of 6 ea | packaging case of 6 ea | packaging case of 4 ea | packaging case of 2 ea |
Still not finding the right product?
Explore all of our products under Nunc® EasyFill Cell Factory System
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?