Przejdź do
TMO132920
Nunc® TripleFlasks, Nunclon® Δ Surface
Nunclon® Delta, surface area (500 cm2), filter cap, straight neck, sterile, case of 32 ea
Synonim(y):
cell culture flask, tissue culture flask, Nunc 132920, surface treated beaker, surface treated flask
Wybierz wielkość
Zmień widok
Do Państwa/SKUDostępnośćCena netto
32 ea
Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności
4750,00 zł
Informacje o tej pozycji
NACRES:
NB.13
UNSPSC Code:
41161502
Material:
screw top blue high-density polyethylene cap, straight neck , straight neck transparent polystyrene flask
4750,00 zł
Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności
Pomoc techniczna
Pozwól nam pomóc
material
screw top blue high-density polyethylene cap, straight neck , straight neck transparent polystyrene flask
sterility
sterile
packaging
case of 32 ea
manufacturer/tradename
Nunc 132920
flask working volume
200 mL
surface area
(500 cm2) , 500 cm2 , culture area (cm2) per flask
working volume
200 mL
binding type
Nunclon Delta surface
General description
Perform cell culture expansion and experience 500cm2 total growth area within the footprint of a standard flask using the Nunc® TripleFlask™ Treated Cell Culture Flasks. Total culture area is achieved by way of the three parallel layers and a straight neck design that works effectively within automation systems. TripleFlask receives the Nunclon™ Delta treatment, a proprietary cell culture surface treatment that offers maximum adhesion for a broad range of cell types. Three layers of growth surfaces provide a total culture area of 500 cm2 in the same footprint as a standard Nunc 175cm2 flask.
Features and Benefits
  • Three parallel growth surfaces provide a total culture area of 500 cm2
  • External dimensions of a 175 cm2 standard flask
  • Ideal for scale-up cell culture applications
  • Extra caps in every carton
  • Nunclon™ Δ surface ensures consistent optimal cell attachment and growth
  • Barcoded with large Code 128
Legal Information
Nunc is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Nunclon is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Porównaj podobne pozycje
Wyświetl pełne porównanie
Pokaż różnice
1 of 1
Ta pozycja
F8667F7552F8542
description
Nunclon® Delta, surface area (500 cm2), filter cap, straight neck, sterile, case of 32 ea
description
Culture flask, surface area 500 cm2, with cap filtered, straight neck, sterile, case of 32 ea
description
Nunclon® Δ surface treatment, 75 cm2 angled neck, filter cap, 100/cs
description
Culture flask, surface area 500 cm2, with solid cap, straight neck, sterile, case of 32 ea
material
straight neck , screw top blue high-density polyethylene cap, straight neck transparent polystyrene flask
material
straight neck , straight neck transparent polystyrene flask, screw top blue high-density polyethylene cap
material
canted neck transparent polystyrene flask, screw top blue high-density polyethylene cap (filter)
material
screw top blue high-density polyethylene cap, straight neck , straight neck transparent polystyrene flask
binding type
Nunclon Delta surface
binding type
Nunclon Delta surface
binding type
Nunclon Delta surface
binding type
Nunclon Delta surface
manufacturer/tradename
Nunc 132920
manufacturer/tradename
Nunc 132913
manufacturer/tradename
Nunc 156499
manufacturer/tradename
Nunc 132867
flask working volume
200 mL
flask working volume
200 mL
flask working volume
25 mL
flask working volume
200 mL
surface area
500 cm2 , culture area (cm2) per flask, (500 cm2)
surface area
500 cm2 , 500 cm2 , culture area (cm2) per flask
surface area
75 cm2
surface area
500 cm2 , culture area (cm2) per flask, 500 cm2
working volume
200 mL
working volume
200 mL
working volume
-
working volume
200 mL
Still not finding the right product?
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)