Wybierz wielkość
| Do Państwa/SKU | Dostępność | Cena netto |
|---|---|---|
10 ea | Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności | 1020,00 zł |
Informacje o tej pozycji
1020,00 zł
material
cap (PP sealing cap), round bottom tube, transparent polycarbonate
sterility
non-sterile
feature
RCF 50,000 × g
packaging
pack of 10
manufacturer/tradename
Nalgene® 3138-0016
nominal capacity
16 mL
General description
Packaging
Legal Information
1 of 1
Ta pozycja | |||
|---|---|---|---|
| description nominal capacity 16 mL | description nominal capacity 30 mL | description nominal capacity 50 mL | description nominal capacity 10 mL |
| manufacturer/tradename Nalgene® 3138-0016 | manufacturer/tradename Nalgene® 3138-0030 | manufacturer/tradename Nalgene® 3138-0050 | manufacturer/tradename Nalgene® 3138-0010 |
| sterility non-sterile | sterility non-sterile | sterility non-sterile | sterility non-sterile |
| material cap (PP sealing cap), transparent polycarbonate , round bottom tube | material cap (PP sealing cap), round bottom tube, transparent polycarbonate | material cap (PP sealing cap), round bottom tube, transparent polycarbonate | material cap (Sealing cap), round bottom tube, transparent polycarbonate |
| packaging pack of 10 | packaging pack of 10 ea | packaging pack of 10 ea | packaging pack of 10 ea |
| feature RCF 50,000 × g | feature RCF 50,000 × g | feature RCF 50,000 × g | feature RCF 50,000 × g |
| nominal capacity 16 mL | nominal capacity 30 mL | nominal capacity 50 mL | nominal capacity 10 mL |
Still not finding the right product?
Explore all of our products under Nalgene® centrifuge tubes, Oak Ridge Style 3138
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?