Informacje o tej pozycji
NACRES:
NB.22
UNSPSC Code:
41122107
Material:
polystyrene
Size:
8 wells , strip
Sterility:
non-sterile
Binding type:
high binding surface
Feature:
lid: no
material
polystyrene
sterility
non-sterile
feature
lid: no
packaging
pack of 1 ea
manufacturer/tradename
Greiner 762061
maximum volume
388 μL
size
8 wells , strip
working volume
350 μL
binding type
high binding surface
General description
Both medium-binding and high binding are offered. High binding plates and strips are recommended for ELISAs. High binding denotes a binding capacity of 600 ng/cm2 and scatter of less than 10%. Optimal protein binding is variable and comparative tests between high and medium binding are suggested.
Legal Information
1 of 1
Ta pozycja | |
|---|---|
| sterility non-sterile | sterility sterile |
| manufacturer/tradename Greiner 762061 | manufacturer/tradename Greiner 781097 |
| feature lid: no | feature lid: no |
| binding type high binding surface | binding type high binding surface |
| material polystyrene | material black polystyrene , flat bottom clear polystyrene wells (with micro-clear bottom) |
| working volume 350 μL | working volume 110 μL |
Still not finding the right product?
Explore all of our products under Greiner Multiwell Strips
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?