Informacje o tej pozycji
NACRES:
NB.01
UNSPSC Code:
41121600
Sterility:
non-sterile
Feature:
barrier: no
Packaging:
pack of 1000 ea (Bag)
Manufacturer/tradename:
Finn 9400230
Pomoc techniczna
Pozwól nam pomócmaterial
colorless
sterility
non-sterile
feature
barrier: no
packaging
pack of 1000 ea (Bag)
manufacturer/tradename
Finn 9400230
maximum volume
250 μL
volume range
0.5-250 μL
Other Notes
Browse all pipette tips
Legal Information
Finnpipette is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Still not finding the right product?
Explore all of our products under Finnpipette® pipette tips
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?