Informacje o tej pozycji
NACRES:
NB.01
UNSPSC Code:
41121500
Manufacturer/tradename:
Drummond Scientific 2-000-040
Material:
glass
Packaging:
pack of 250 ea
material
glass
packaging
pack of 250 ea
manufacturer/tradename
Drummond Scientific 2-000-040
measuring range
accuracy: ±0.5%
capacity
40 μL
color
color-coded yellow
General description
- Drummond Calibrated Micropipettes provide accuracy and convenience for a wide variety of liquid transfer applications.
- Made of high quality glass.
- Fire polished micropipettes are imprinted with volume and a color-code bar for easy identification.
- Calibrated "To Deliver."
High quality Drummond micropipettes are fire polished, imprinted with volume and a color-code bar for easy identification. Calibrated "To Deliver."
1 of 1
Ta pozycja | |||
|---|---|---|---|
| material glass | material glass | material glass | material glass |
| manufacturer/tradename Drummond Scientific 2-000-040 | manufacturer/tradename Drummond Scientific 2-000-010 | manufacturer/tradename Drummond Scientific 2-000-100 | manufacturer/tradename Drummond Scientific 2-000-020 |
| packaging pack of 250 ea | packaging pack of 250 ea | packaging pack of 250 ea | packaging pack of 250 ea |
| color color-coded yellow | color color-coded orange | color color-coded blue | color color-coded black |
| capacity 40 μL | capacity 10 μL | capacity 100 μL | capacity 20 μL |
| measuring range accuracy: ±0.5% | measuring range accuracy: ±0.5% | measuring range accuracy: ±0.5% | measuring range accuracy: ±0.5% |
Still not finding the right product?
Explore all of our products under Microcapillary tube, calibrated
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?