Merck
OH83033966
Ohaus® pH Electrode
ST210
Informacje o tej pozycji
UNSPSC Code:
41115604
Pomoc techniczna
Pozwól nam pomóc
packaging
pack of 1 ea
manufacturer/tradename
Ohaus® 83033966
pH measuring range
0.00-14 pH unit
compatibility
all Ohaus Starter bench and portable meters
General description
All electrodes include a temperature sensor, which powers automatic temperature compensation and ensures accurate measurements.
Application
pH, reference, oxidation-reduction potential (ORP), conductivity, dissolved oxygen (DO), temperature measurement
Features and Benefits
Can be used in conjunction with all Starter bench and portable meters
Legal Information
Ohaus is a registered trademark of Ohaus Corp.
Still not finding the right product?
Explore all of our products under Ohaus® pH Electrode
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)