Przejdź do
HS120593
Premium Plus Slide Box
to hold, 100 slide(s), translucent assorted colors acrylonitrile-butadiene-styrene (ABS)
Wybierz wielkość
Zmień widok
Do Państwa/SKUDostępnośćCena netto
5 ea
Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności
371,00 zł
Informacje o tej pozycji
NACRES:
NB.16
UNSPSC Code:
41122800
Manufacturer/tradename:
Heathrow Scientific 120593
Feature:
to hold 100 slide(s)
Material:
translucent assorted colors acrylonitrile-butadiene-styrene (ABS)
Packaging:
pack of 1 ea
371,00 zł
Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności
Pomoc techniczna
Pozwól nam pomóc
material
translucent assorted colors acrylonitrile-butadiene-styrene (ABS)
feature
to hold 100 slide(s)
packaging
pack of 1 ea
manufacturer/tradename
Heathrow Scientific 120593
General description
The Premium Plus Slide Box′s modern design for daily use is stylish with a wave surface texture. The slide box is easy to hold,increasing safety of sample transport with an EVA foam lined bottom of the box helping to hold slides securely in placeand protect against damage. Its unique side storage can accommodate a pen, desiccant or other small items needed for working.
Features and Benefits
  • Made from ABS and Polypropylene makes the boxes robust and eliminates potential rusting of metal parts
  • Inventory card is removable and replaceable for trouble-free writing on flat surfaces.
  • Hinged lid smoothly opens 180° sitting flat on surfaces and prevents misplacement
  • EVA foam lines the bottom of the box helping to hold slides securely in place and protect against damage
  • Multiple labeling areas, a large area on the lid of the box with “project” and “date” molded in and the back of the index card can be used as the lid is transparent
  • Vibrant translucent colors are easy to identify and enable coding options
  • Stylish wave surface texture is easy to hold, increasing safety of sample transport
  • Stackable, molded feet allow secure horizontal stacking of multiple units for safe transport or storing
  • Flexible storage, the units can be stored vertically or horizontally helping to efficiently use space available
  • Unique side storage can accommodate a pen, desiccant or other small items needed for working
  • Holds 100 - 26 x 76 mm slides
  • Patent Pending
Porównaj podobne pozycje
Wyświetl pełne porównanie
Pokaż różnice
1 of 1
Ta pozycja
HS120577HS120578HS120576
description
to hold, 100 slide(s), translucent assorted colors acrylonitrile-butadiene-styrene (ABS)
description
to hold, 100 slide(s), translucent orange acrylonitrile-butadiene-styrene (ABS)
description
to hold, 100 slide(s), translucent green acrylonitrile-butadiene-styrene (ABS)
description
to hold, 100 slide(s), translucent blue acrylonitrile-butadiene-styrene (ABS)
manufacturer/tradename
Heathrow Scientific 120593
manufacturer/tradename
Heathrow Scientific 120577
manufacturer/tradename
Heathrow Scientific 120578
manufacturer/tradename
Heathrow Scientific 120576
feature
to hold 100 slide(s)
feature
to hold 100 slide(s)
feature
to hold 100 slide(s)
feature
to hold 100 slide(s)
material
translucent assorted colors acrylonitrile-butadiene-styrene (ABS)
material
polypropylene , translucent orange acrylonitrile-butadiene-styrene (ABS)
material
translucent green acrylonitrile-butadiene-styrene (ABS)
material
polypropylene , translucent blue acrylonitrile-butadiene-styrene (ABS)
packaging
pack of 1 ea
packaging
pack of 1 ea
packaging
pack of HS120578
packaging
pack of 5 ea
Still not finding the right product?
Explore all of our products under Premium Plus Slide Box
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)