Wybierz wielkość
| Do Państwa/SKU | Dostępność | Cena netto |
|---|---|---|
1 ea | Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności | 626,00 zł |
Informacje o tej pozycji
626,00 zł
material
natural , polyolefin (Thermoplastic)
sterility
non-sterile
packaging
pack of 1 ea
manufacturer/tradename
Heathrow Scientific HS120265, Parafilm PM998
film W × L
508 mm × 16 m
General description
Features and Benefits
- Protect the integrity of research applications with a tight moisture-barrier that holds a strong seal and doesn′t release from glassware, despite off-gassing
- Reduce the risk of tampered data with a covering that is odorless, colorless and semi-transparent
- Prevents moisture and volume loss due to material composition and tight seal
- Flexible and self-sealing material allows it to cling around irregular shapes and surfaces
- Stretches more than 200% of original length
- Offers good resistance against acids, alkalis and some organic solvents
- Resistant up to 48 hours against many polar substances such as saline solutions, inorganic acids and alkaline solutions
- Disposable
Legal Information
1 of 1
Ta pozycja | |||
|---|---|---|---|
| sterility non-sterile | sterility non-sterile | sterility non-sterile | sterility non-sterile |
| manufacturer/tradename Heathrow Scientific HS120265, Parafilm PM998 | manufacturer/tradename Heathrow Scientific HS234526A, Parafilm PM992 | manufacturer/tradename Heathrow Scientific HS234526B, Parafilm PM996 | manufacturer/tradename Heathrow Scientific HS234526C, Parafilm PM999 |
| material natural , polyolefin (Thermoplastic) | material natural , polyolefin (Thermoplastic) | material natural , polyolefin (Thermoplastic) | material natural , polyolefin (Thermoplastic) |
| packaging pack of 1 ea | packaging pack of 1 ea | packaging pack of 1 ea | packaging pack of 1 ea |
| film W × L 508 mm × 16 m | film W × L 50 mm × 75 m | film W × L 100 mm × 38 m | film W × L 100 mm × 75 m |
Still not finding the right product?
Explore all of our products under Parafilm® M Sealing Film
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?