Wybierz wielkość
| Rozmiar/SKU | Dostępność | Cena netto |
|---|---|---|
1 ea | Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności | 174,00 zł 147,90 zł |
Informacje o tej pozycji
147,90 zł
Cena katalogowa174,00 złZaoszczędź 15%material
polypropylene cap, yellow PE-LD bottle
joint
joint threaded
feature
label no label, vented, with cap
packaging
pack of 6 ea
manufacturer/tradename
Heathrow Scientific HS120247
capacity
500 mL
General description
The closure and stem are molded in one piece to make them leak-proof. The angled neck and tip provide a uniform stream of liquid and the tip can be cut back if required to increase the flow.
Application
Features and Benefits
- Wide bottle mouth makes filling easier
- Color-coding for user, application or location identification
- The closure and stem are molded in one piece to make them leak-proof
- Bottle is made from LDPE, with a PP closure
- 500 mL bottle size
- Supplied in packs of 6
1 of 1
Ta pozycja | |||
|---|---|---|---|
| description capacity 500 mL, yellow PE-LD bottle | description white PE-LD bottle, capacity 500 mL | description capacity 500 mL, red PE-LD bottle | description capacity 500 mL, blue PE-LD bottle |
| capacity 500 mL | capacity 500 mL | capacity 500 mL | capacity 500 mL |
| feature vented, label no label, with cap | feature vented, with cap, label no label | feature with cap, label no label, vented | feature vented, with cap, label no label |
| manufacturer/tradename Heathrow Scientific HS120247 | manufacturer/tradename Heathrow Scientific HS120245 | manufacturer/tradename Heathrow Scientific HS120246 | manufacturer/tradename Heathrow Scientific HS120249 |
| material yellow PE-LD bottle, polypropylene cap | material white PE-LD bottle, polypropylene cap | material red PE-LD bottle, polypropylene cap | material polypropylene cap, blue PE-LD bottle |
| joint joint threaded | joint joint threaded | joint joint threaded | joint joint threaded |
| packaging pack of 6 ea | packaging pack of 6 ea | packaging pack of 6 ea | packaging pack of 6 ea |
Still not finding the right product?
Explore all of our products under Wide mouth wash bottle
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?