Wybierz wielkość
| Do Państwa/SKU | Dostępność | Cena netto |
|---|---|---|
500 ea | Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności | 508,00 zł |
Informacje o tej pozycji
508,00 zł
material
white polystyrene (Anti-static)
packaging
pk of 500
manufacturer/tradename
Heathrow Scientific HS120226
L × W × H
83 mm × 132 mm × 26 mm
General description
Features and Benefits
- biologically inert and resistant to dilute and weak acids, aqueous solutions, alcohols, and bases.
- temperature range -10 to 70C
- disposable
- pour-spout design easily dispenses samples without flexing the dish
- flat-bottom dishes are made from 12 mil think polystyrene for better strength, stability, and handling
1 of 1
Ta pozycja | |||
|---|---|---|---|
| description medium, white polystyrene (Anti-static), pk of 500 | description small, white polystyrene (Anti-static), pk of 500 | description large, white polystyrene (Anti-static), pk of 500 | description medium, white polystyrene, pack of 500 ea |
| manufacturer/tradename Heathrow Scientific HS120226 | manufacturer/tradename Heathrow Scientific HS120225 | manufacturer/tradename Heathrow Scientific HS120227 | manufacturer/tradename Heathrow Scientific HS1419B |
| material white polystyrene (Anti-static) | material white polystyrene (Anti-static) | material white polystyrene (Anti-static) | material white polystyrene |
| packaging pk of 500 | packaging pk of 500 | packaging pk of 500 | packaging pack of 500 ea |
| L × W × H 83 mm × 132 mm × 26 mm | L × W × H 43 mm × 58 mm × 13 mm | L × W × H 108 mm × 183 mm × 26 mm | L × W × H 83 mm × 132 mm × 26 mm |
Still not finding the right product?
Explore all of our products under Pour Boat Weighing Dishes
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?