Przejdź do
GBLRD476687
Grace Bio-Labs wound splint, w/ suture sites
red silicone, O.D. × I.D. × thickness 14 mm × 7 mm × 0.5 mm
Synonim(y):
silicone wound splints, suture site template, Grace Biolabs 476687, wound split device, wound suture template
Wybierz wielkość
Zmień widok
Rozmiar/SKUDostępnośćCena netto
Informacje o tej pozycji
NACRES:
NB.22
UNSPSC Code:
41122600
Material:
red silicone
Packaging:
pkg of 20 ea (2 splints per pouch)
Manufacturer/tradename:
Grace Bio-Labs 476687
Informacje o cenach i dostępności nie są obecnie dostępne.
Pomoc techniczna
Pozwól nam pomóc
material
red silicone
sterility
(sterile)
packaging
pkg of 20 ea (2 splints per pouch)
manufacturer/tradename
Grace Bio-Labs 476687
O.D. × I.D. × thickness
14 mm × 7 mm × 0.5 mm
General description
Grace Bio-Labs has worked with researchers to develop and manufacture wound splint devices that aid in the understanding of wound and tissue repair for animal research and the testing of new therapies for safety and efficacy.
Wound splints are constructed of silicone and include suture sites for increased precision in affixing on or within. These devices enable reproducible and reliable experimental conditions for wound healing in animal research.
The splints are offered in a variety of shapes and sizes (visible stitch holes / suture sites, 5 mm dia.), shipped sterile and ready to use.
Porównaj podobne pozycje
Wyświetl pełne porównanie
Pokaż różnice
1 of 1
Ta pozycja
GBLRD476685GBLRD476684GBL664384
description
red silicone, O.D. × I.D. × thickness 14 mm × 7 mm × 0.5 mm
description
clear silicone, O.D. × I.D. × thickness 24 mm × 20 mm × 0.5 mm
description
clear silicone, O.D. × I.D. × thickness 14 mm × 10 mm × 0.5 mm
description
thickness 2.4 mm, (No adhesive)
manufacturer/tradename
Grace Bio-Labs 476687
manufacturer/tradename
Grace Bio-Labs 476685
manufacturer/tradename
Grace Bio-Labs 476684
manufacturer/tradename
Grace Bio-Labs 664384
material
red silicone
material
clear silicone
material
clear silicone
material
red silicon
packaging
pkg of 20 ea (2 splints per pouch)
packaging
pkg of 20 ea (2 splints per pouch)
packaging
pkg of 20 ea (2 splints per pouch)
packaging
pack of 5 ea
sterility
(sterile)
sterility
(sterile)
sterility
(sterile)
sterility
non-sterile
O.D. × I.D. × thickness
14 mm × 7 mm × 0.5 mm
O.D. × I.D. × thickness
24 mm × 20 mm × 0.5 mm
O.D. × I.D. × thickness
14 mm × 10 mm × 0.5 mm
O.D. × I.D. × thickness
-
Still not finding the right product?
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)