Zmień widok
| Rozmiar/SKU | Dostępność | Cena netto |
|---|---|---|
5 ea | Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności | 423,00 zł 359,55 zł |
Informacje o tej pozycji
NACRES:
NB.15
UNSPSC Code:
41122410
eCl@ss:
32120613
Manufacturer/tradename:
Grace Bio-Labs 845236
Material:
paper film
W × l:
78 mm × 120 mm
Sterility:
non-sterile
Packaging:
pack of 5 ea
359,55 zł
Cena katalogowa423,00 złZaoszczędź 15%Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności
Pomoc techniczna
Pozwól nam pomócmaterial
paper film
sterility
non-sterile
feature
wells 96
packaging
pack of 5 ea
manufacturer/tradename
Grace Bio-Labs 845236
W × L
78 mm × 120 mm
General description
ProCrystal protein crystallization covers are designed for high throughput protein x-ray crystallography in 96-well plates. The ProCrystal material is extremely hydrophobic to maintain drop footprint and segregation, even with protocols using MPD, glycerol or detergents. Grace Bio-Labs has collaborated with leading crystallographers to develop our ProCrystal covers using materials with the highest optical quality and stringent manufacturing processes to ensure clean, particle-free covers.
Features and Benefits
- TTP Labtech Mosquito compatible
- Extremely hydrophobic
- Ready-to-use, dust-free
- UV compatible
- No noticeable x-ray diffraction
- Accommodates up to 3 protein drops per well
- Two formats: pre-cut or uncut
Legal Information
ProCrystal is a trademark of Grace Bio-Labs, Inc.
1 of 1
Ta pozycja | |||
|---|---|---|---|
| description cut per individual well, paper liner, wells, 96 | description uncut, plastic liner | description cut per individual well, plastic liner, wells, 96 | description 24 mm x 77 mm seal area, non-sterile |
| sterility non-sterile | sterility non-sterile | sterility non-sterile | sterility non-sterile |
| W × L 78 mm × 120 mm | W × L 78 mm × 120 mm | W × L 78 mm × 120 mm | W × L - |
| manufacturer/tradename Grace Bio-Labs 845236 | manufacturer/tradename Grace Bio-Labs 845233 | manufacturer/tradename Grace Bio-Labs 845237 | manufacturer/tradename Grace Bio-Labs 204974 |
| material paper film | material plastic film | material plastic film | material polyolefin |
| packaging pack of 5 ea | packaging pack of 5 ea | packaging pack of 5 ea | packaging pack of 50 ea |
| feature wells 96 | feature - | feature wells 96 | feature - |
Still not finding the right product?
Explore all of our products under Grace Bio-Labs ProCrystal™ protein crystallization cover
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?