Zmień widok
| Rozmiar/SKU | Dostępność | Cena netto |
|---|
Informacje o tej pozycji
UNSPSC Code:
41121813
Aby zapytać o ten produkt EP952010077, skontaktuj się z lokalnym biurem lub sprzedawcą firmy Merck. Skontaktuj się z zespołem ds. pomocy technicznej
Pomoc techniczna
Pozwól nam pomócmaterial
(UV-transparent plastic)
packaging
pack of 1
manufacturer/tradename
Eppendorf® 952010077
General description
UVette® starter set, 80 UVettes and 1 universal adapter for light path height 15 mm, convertible to 8.5 mm, 50 – 2,000 µL
- individually packaged
- PCR-clean and protein-free quality
- UV transparent plastic, free of fluoropolymers and other halogenated hydrocarbons
- Two built-in optical path lengths in a single cuvette - just turn the UVette 90° to change from 10 mm and 2 mm
- Volume markings for 500 µL and 1,000 µL
- Individually packaged and certified PCR clean and protein-free quality for sensitive (e.g. RNA) and valuable samples
- Deep-lying optical window prevents scratches
- Tapered cuvette base for optimal filling, frosted grip for labeling
- Ideal for use in the Eppendorf BioPhotometer and Eppendorf BioSpectrometer, adapters available for use with other common spectrophotometers
- Routine pack for convenient access to each UVette and safe storage in a reclosable box
Features and Benefits
- Individually packaged
- PCR-clean and protein-free quality
- UV transparent plastic, free of fluoropolymers and other halogenated hydrocarbons
- Two built-in optical path lengths in a single cuvette - just turn the UVette 90° to change from 10 mm and 2 mm
- Volume markings for 500 µL and 1,000 µL
- Individually packaged and certified PCR clean and protein-free quality for sensitive (e.g. RNA) and valuable samples
- Deep-lying optical window prevents scratches
- Tapered cuvette base for optimal filling, frosted grip for labeling
- Ideal for use in the Eppendorf BioPhotometer and Eppendorf BioSpectrometer, adapters available for use with other common spectrophotometers
- Routine pack for convenient access to each UVette and safe storage in a reclosable box
Legal Information
Eppendorf is a registered trademark of Eppendorf SE
UVette is a registered trademark of Eppendorf SE
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?