Wybierz wielkość
| Rozmiar/SKU | Dostępność | Cena netto |
|---|---|---|
1 ea | Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności | 1980,00 zł |
Informacje o tej pozycji
1980,00 zł
feature
to hold 70 x 1.5/2.0 mL reaction vessels, fixed angle rotor, non aerosol tight
packaging
pack of 1 ea
manufacturer/tradename
Eppendorf® 5490032004 (EU)
technique(s)
centrifugation: suitable
compatibility
for use with centrifuge models Concentrator Plus
General description
Legal Information
1 of 1
Ta pozycja | |||
|---|---|---|---|
| description F-45-70-11, non aerosol tight, to hold, 70 x 1.5/2.0 mL reaction vessels | description F-40-18-19, non aerosol tight, to hold, 18 x 10 mL flat-bottom tubes | description F-45-16-20, non aerosol tight, to hold, 16 x 6.5/10 mL flat-bottom tubes | description F-45-36-15, non aerosol tight, to hold, 36 x 3.0/5.0 mL flat-bottom tubes |
| compatibility for use with centrifuge models Concentrator Plus | compatibility for use with centrifuge models Concentrator Plus | compatibility for use with centrifuge models Concentrator Plus | compatibility for use with centrifuge models Concentrator Plus |
| manufacturer/tradename Eppendorf® 5490032004 (EU) | manufacturer/tradename Eppendorf® 5490037006 (EU) | manufacturer/tradename Eppendorf® 5490043006 (EU) | manufacturer/tradename Eppendorf® 5490035003 (EU) |
| feature to hold 70 x 1.5/2.0 mL reaction vessels, non aerosol tight, fixed angle rotor | feature to hold 18 x 10 mL flat-bottom tubes, to hold 18 x 19 x 66 mm, non aerosol tight, fixed angle rotor | feature to hold 16 x 20 x 42 - 55 mm, fixed angle rotor, non aerosol tight, to hold 16 x 6.5/10 mL flat-bottom tubes | feature fixed angle rotor, to hold 36 x 15 x 48 mm, non aerosol tight, to hold 36 x 3.0/5.0 mL flat-bottom tubes |
| packaging pack of 1 ea | packaging pack of 1 ea | packaging pack of 1 ea | packaging pack of 1 ea |
| technique(s) centrifugation: suitable | technique(s) centrifugation: suitable | technique(s) centrifugation: suitable | technique(s) centrifugation: suitable |
rotor options for Eppendorf Concentrator Plus | Centrifuge rotor compatible with Eppendorf Concentrator Plus, Eppendorf Concentrator Plus replacement rotor, Eppendorf Concentrator Plus rotor specifications, Eppendorf rotor for sample concentration, high-performance rotor for Eppendorf Concentrator Plus, laboratory centrifuge rotor for Eppendorf, Eppendorf Concentrator Plus rotor for laboratory applications, centrifugation rotor for Eppendorf Concentrator Plus, Eppendorf rotor accessory for Concentrator Plus, Eppendorf Concentrator Plus rotors, Eppendorf Concentrator Plus rotor options, Eppendorf centrifuge rotor for Concentrator Plus, Eppendorf Concentrator Plus centrifugation rotor, Eppendorf Concentrator Plus rotor selection, custom Eppendorf Concentrator Plus rotors | centrifugation rotor for Eppendorf Concentrator Plus, Eppendorf rotor accessories for Concentrator Plus, Eppendorf Concentrator Plus rotor specifications, Eppendorf rotor for high-throughput centrifugation, Eppendorf Concentrator Plus rotor for laboratory use, Eppendorf Concentrator Plus rotors, high-performance rotor for Eppendorf Concentrator Plus, compatible rotor for Eppendorf Concentrator Plus centrifuge, centrifuge rotor selection for Eppendorf Concentrator Plus, Eppendorf Concentrator Plus centrifuge rotor options, Eppendorf rotor replacement for Concentrator Plus, Eppendorf Concentrator Plus rotor parts | - |
Still not finding the right product?
Explore all of our products under Rotor for Eppendorf® Concentrator Plus
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?