Merck
EP0030129334
Eppendorf® Twin tec microbiology PCR plates, 96 wells
blue plate, 96, semi-skirted, conical bottom colorless wells, PCR clean, semi-skirted, pkg of 10 ea, blue
Informacje o tej pozycji
NACRES:
NC.22
UNSPSC Code:
41121800
Material:
blue plate, conical bottom colorless wells
Size:
96 wells
Sterility:
non-sterile
Feature:
PCR clean, skirt, 96, semi-skirted
Pomoc techniczna
Pozwól nam pomóc
material
blue plate, conical bottom colorless wells
sterility
non-sterile
feature
PCR clean, skirt, 96, semi-skirted
packaging
pkg of 10 ea
manufacturer/tradename
Eppendorf® 0030129334
size
96 wells
color
blue
Features and Benefits
  • Individually blistered Certified PCR clean
  • Additionally free of bacterial DNA
  • For all applications that need ultimate purity or certified absense of bacterial DNA
  • Ideal for quantitative real-time PCR as well – OptiTrack matrix for faster sample identification and fewer pipetting errors
Legal Information
Eppendorf is a registered trademark of Eppendorf SE
Still not finding the right product?
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)