Przejdź do
CLS6633
Corning® Step-R Repeating Pipettor Syringe Tips
volume (12.5 mL), sterile
Synonim(y):
pipet tips
Informacje o tej pozycji
NACRES:
NB.02
UNSPSC Code:
41121600
Volume:
(12.5 mL)
Sterility:
sterile
Packaging:
pack of 100 ea
Manufacturer/tradename:
Corning 6633
sterility
sterile
packaging
pack of 100 ea
manufacturer/tradename
Corning 6633
volume
(12.5 mL)
General description
  • Ideal for viscous, dense and volatile liquides
  • Minimum increments printed on the syringe itself
  • Accurately deliver selected dose
  • Made from high quality polypropylene and polyethylene
Legal Information
Corning is a registered trademark of Corning, Inc.
Porównaj podobne pozycje
Wyświetl pełne porównanie
Pokaż różnice
1 of 1
Ta pozycja
CLS6625CLS6622CLS6631
description
volume (12.5 mL), sterile
description
volume (12.5 mL), non-sterile
description
volume (1.25 mL), non-sterile
description
volume (2.5 mL), sterile
sterility
sterile
sterility
non-sterile
sterility
non-sterile
sterility
sterile
volume
(12.5 mL)
volume
(12.5 mL)
volume
(1.25 mL)
volume
(2.5 mL)
manufacturer/tradename
Corning 6633
manufacturer/tradename
Corning 6625
manufacturer/tradename
Corning 6622
manufacturer/tradename
Corning 6631
packaging
pack of 100 ea
packaging
pack of 100 ea
packaging
pack of 100 ea
packaging
pack of 100 ea
Still not finding the right product?
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)