Informacje o tej pozycji
NACRES:
NB.11
UNSPSC Code:
41121522
Manufacturer/tradename:
Corning 4877
Capacity:
200 μL , 5 mL
Material:
U-bottom bottom, white polystyrene
Sterility:
sterile
Pomoc techniczna
Pozwól nam pomócmaterial
U-bottom bottom, white polystyrene
sterility
sterile
packaging
pack of 100 ea
manufacturer/tradename
Corning 4877
volume measuring range
25-200 μL
capacity
200 μL , 5 mL
volume
200 μL
General description
The Transtar is a portable, autoclavable liquid handling system designed for use with 96 well plates, A sterile 96 tip disposable cartridge, which loads into the Transtar system, enables liquids to be aspirated, transferred and dispensed over a volume range of 25 - 200 μL in 5 μL increments, Transtar accuracy us rated at 5% at all volume levels.
Legal Information
Corning is a registered trademark of Corning, Inc.
TRANSTAR-96 is a registered trademark of Corning, Inc.
Still not finding the right product?
Explore all of our products under Corning® Transtar® liquid handling system
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?