Wybierz wielkość
| Do Państwa/SKU | Dostępność | Cena netto |
|---|---|---|
20 ea | Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności | 2620,00 zł |
Informacje o tej pozycji
2620,00 zł
material
black cyclic olefin copolymer (COC) , clear bottom, flat bottom , flat bottom wells
sterility
sterile
feature
low base, lid, film bottom
packaging
pack of 20 plates, pack of 20
manufacturer/tradename
Corning 4517
surface area
0.16 cm2 , cell growth area
wall volume
190 μL
wall working volume
50 μL
suitability
suitable for (cell cuture)
Legal Information
1 of 1
Ta pozycja | |||
|---|---|---|---|
| suitability suitable for (cell cuture) | suitability suitable for (cell cuture) | suitability suitable for (cell cuture) | suitability suitable for (cell cuture) |
| material black cyclic olefin copolymer (COC) , flat bottom , clear bottom, flat bottom wells | material black cyclic olefin copolymer (COC) , clear bottom, flat bottom , flat bottom wells | material black wells (with 0.2 mm glass bottom), clear bottom, flat bottom , flat bottom wells | material black cyclic olefin copolymer (COC) (COC), clear bottom, flat bottom |
| feature lid, low base, film bottom | feature lid, film bottom, low base | feature lid, skirt, glass bottom | feature lid, skirt, black cyclic olefin copolymer (COC), half area, HCS, film bottom |
| packaging pack of 20 plates | packaging pack of 20 plates | packaging pack of 10, pack of 20 plates | packaging pack of 20 |
| surface area 0.16 cm2 , cell growth area | surface area 0.03 cm2 , cell growth area | surface area - | surface area - |
| manufacturer/tradename Corning 4517 | manufacturer/tradename Corning 4518 | manufacturer/tradename Corning 4581 | manufacturer/tradename Corning 4681 |
Still not finding the right product?
Explore all of our products under Corning® High Content Imaging Microplate
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?