Przejdź do
CLS3925
Corning® 96 well plates
flat bottom, black polystyrene, high binding surface, bag of 25, non-sterile, lid: no
Synonim(y):
96 microtiter plates, 96 multiwell plates, 96 well microplates, 96 well plates
Wybierz wielkość
Zmień widok
Rozmiar/SKUDostępnośćCena netto
100 ea
Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności
2780,00 zł
Informacje o tej pozycji
NACRES:
NB.13
UNSPSC Code:
41121800
Material:
black bottom, black polystyrene , flat bottom , polystyrene
Size:
96 wells
2780,00 zł
Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności
​
Pomoc techniczna
Pozwól nam pomóc
material
black bottom, black polystyrene , flat bottom , polystyrene
sterility
non-sterile
feature
lid: no, skirt, plate format: 96 well standard
packaging
case of 100, bag of 25, pack of 25
manufacturer/tradename
Corning 3925
size
96 wells
well volume
360 μL
well working volume
75-200 μL
suitability
suitable for (cell cuture)
binding type
high binding surface
General description
  • Black microplates have low background fluorescence and minimize light scattering
  • Designed to reduce well-to-well crosstalk
  • High binding surface is capable of binding medium (>10kD) and large biomolecules that possess ionic groups and/or hydrophobic regions
  • Binding capacity is approximately 400 to 500ng IgG/cm2
  • Ideal for immunoassays
  • Without lids, lids supplied separately
  • Total volume is 360μL; recommended working volume of 75 to 200μL
  • Conforms to standard microplate footprint and dimensions
Legal Information
Corning is a registered trademark of Corning, Inc.
Porównaj podobne pozycje
Wyświetl pełne porównanie
Pokaż różnice
1 of 1
Ta pozycja
CLS3922CLS3340CLS3641
description
flat bottom, black polystyrene, high binding surface, bag of 25, non-sterile, lid: no
description
96 well plate, polystyrene, High Bind, white flat bottom wells, non-sterile, white, 100/cs
description
flat bottom clear, black polystyrene, sterile, lid
description
flat bottom clear, polystyrene, bag of 25 ×, non-sterile, lid: no
suitability
suitable for (cell cuture)
suitability
suitable for (cell cuture)
suitability
suitable for (cell cuture)
suitability
suitable for (cell cuture)
size
96 wells
size
96 wells
size
96 wells
size
96 wells
material
flat bottom , polystyrene , black polystyrene , black bottom
material
white polystyrene plate, flat bottom wells white
material
flat bottom wells , round wells, flat bottom clear , black plate, black polystyrene , polystyrene plate
material
polystyrene , round clear wells, flat bottom clear
feature
skirt, plate format: 96 well standard, lid: no
feature
plate format: 96 well standard, lid: no, skirt
feature
lid, plate format: 96 well standard, skirt
feature
lid: no, skirt, plate format: 96 well standard
packaging
bag of 25, pack of 25, case of 100
packaging
case of 100, pack of 25
packaging
case of 50, pack of 5
packaging
case of 100, pack of 25, bag of 25 ×
sterility
non-sterile
sterility
non-sterile
sterility
sterile
sterility
non-sterile
Still not finding the right product?
Explore all of our products under Corning® 96 well plates
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)