Przejdź do
CLS3578
Corning® disposable spinner flasks
disposable spinner flask, polystyrene, non-treated, 500 mL, w/ vent caps, sterile, 12/cs
Synonim(y):
cell culture flasks, cell culture spinner flasks, scale up cell culture flasks
, aby wyświetlić ceny organizacyjne i kontraktowe.
Wybierz wielkość
Zmień widok
Do Państwa/SKUDostępnośćCena netto
1 ea
Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności
843,00 zł
12 ea
Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności
7260,00 zł
Informacje o tej pozycji
NACRES:
NB.43
UNSPSC Code:
41121804
Capacity:
500 mL
Material:
polyethylene cap, polystyrene flask
Sterility:
sterile
Packaging:
case of 12
Feature:
w/ vent caps
843,00 zł
Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności
Pomoc techniczna
Pozwól nam pomóc
material
polyethylene cap, polystyrene flask
sterility
sterile
feature
w/ vent caps
packaging
case of 12
manufacturer/tradename
Corning 3578
capacity
500 mL
working volume
300 mL
fitting
cap for GL 45, GL 45 neck
suitability
suitable for (scale-up cell culture; production of secreted proteins)
General description
Convenience
The Corning disposable spinner flask system comes ready-to-use with paddle and integrated magnet, eliminating the need for time-consuming assembly or cleaning and reassembly.
Confidence
Molded from virgin polystyrene and gamma-irradiated, each spinner flask system assures a clean and sterile unit that is nonpyrogenic. No more concerns with detergent residues or contamination.
Compatibility
Made of ISO 10993 polystyrene, the vessel is comparable to conventional glass spinner flasks for growth of suspension cell lines and any attachment-dependent cultures using microcarrier beads.
Performance
The paddle size and height is optimized for each vessel size. A unique integrated magnet provides a smooth, even rotation at required speeds on any laboratory slow speed stirrer. Heat build-up in the vessel is reduced by means of a specially designed flange that raises the vessel off the stir-plate surface.
Conclusions
  • Disposable large volume spinner flasks are equivalent to conventional glass spinner flasks for cell growth and viability
  • A variety of cell types can be cultured in large volume disposable spinner flasks
  • Disposable spinner flasks maintain equal aeration as compared to standard glass spinner flasks
  • Cultured cells from glass and disposable spinner flasks are metabolically equivalent
  • Culured cells in disposable spinner flasks product equal amounts of secreted proteins as cells cultured in traditional glass spinner flasks
Corning disposable spinner flasks are designed to be single-use culture flask with no assembly required. These spinner flasks are suitable as scale-up cell culture vessels in addition to the production of secreted proteins.
Legal Information
Corning is a registered trademark of Corning, Inc.
Porównaj podobne pozycje
Wyświetl pełne porównanie
Pokaż różnice
1 of 1
Ta pozycja
CLS3153CLS3580CLS3152
manufacturer/tradename
Corning 3578
manufacturer/tradename
Corning 3153
manufacturer/tradename
Corning 3580
manufacturer/tradename
Corning 3152
sterility
sterile
sterility
sterile
sterility
sterile
sterility
sterile
capacity
500 mL
capacity
500 mL
capacity
1 L
capacity
125 mL
material
polyethylene cap, polystyrene flask
material
polypropylene cap, polystyrene flask, screw top cap
material
high-density polyethylene cap, polystyrene flask, vent cap
material
polypropylene cap, polystyrene flask, screw top cap
packaging
case of 12
packaging
case of 12
packaging
case of 6
packaging
case of 12
fitting
cap for GL 45, GL 45 neck
fitting
-
fitting
cap for GL 45, GL 45 neck
fitting
-
Still not finding the right product?
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)