Informacje o tej pozycji
NACRES:
NB.13
UNSPSC Code:
41122102
Material:
flat clear polystyrene
Size:
96 wells
Sterility:
sterile
Binding type:
cell culture treated
Feature:
lid
Pomoc techniczna
Pozwól nam pomócmaterial
flat clear polystyrene
sterility
sterile
feature
lid
manufacturer/tradename
Corning 353872
technique(s)
cell culture | mammalian: suitable
size
96 wells
binding type
cell culture treated
Legal Information
Corning is a registered trademark of Corning, Inc.
Still not finding the right product?
Explore all of our products under Corning® Primaria Cell Culture Plate
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?