Wybierz wielkość
| Do Państwa/SKU | Dostępność | Cena netto |
|---|---|---|
8 ea | Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności | 9610,00 zł |
Informacje o tej pozycji
9610,00 zł
material
cap (vented), polystyrene , rectangular , rectangular bottom flask, straight neck , vent cap
sterility
sterile (γ-irradiated)
feature
cap, graduations
packaging
pack of 1, case of 8
manufacturer/tradename
Corning 3303
surface area
636 cm2
working volume
125-190 mL
suitability
suitable for (scale-up cell culture)
binding type
low binding surface
General description
- Ideal for cell culture scale-up
- Culture chambers produce cell yields in the 107 - 109 range
- Useful alternatives to multiple roller bottles or spinner flasks.
- High mechanical strength and structural integrity
- 100% leak tested prior to shipping
- Certified nonpyrogenic and sterilized by gamma irradiation, with low particulate contamination from modern manufacturing methods
- Easy to use larger openings with threaded closures and vented caps
- Large 26 mm wide filling ports allow direct access to chamber bottom, with more options for sterile transfer of media and cultures
Features and Benefits
- Cell growth area is 636 cm2.
- Ultra Low Attachment surface features a covalently bound hydrogel layer that effectively inhibits cellular attachment and minimizes protein absorption, enzyme activation, and cellular activation.
- Surface is non-cytotoxic, biologically inert, and nondegradable.
- Two 26 mm diameter filling allow direct access to chamber bottom providing greater flexibility for sterile filling and emptying by pouring, pipetting or via tubing in a fully closed system.
- Standard 33 mm threaded orange caps have 0.2 μm pore nonwettable membranes sealed directly to the caps to allow gas exchange (vented caps) while minimizing the risk of contamination.
- Optional 33 mm threaded caps are available with integrally sealed USP Class VI certified C-Flex tubing to allow direct sterile transfer of media and cells.
- USP Class VI polystyrene provides excellent optical clarity and mechanical strength.
- Sterilized by gamma radiation and certified nonpyrogenic.
- 100% leak tested prior to shipping.
Legal Information
1 of 1
Ta pozycja | |||
|---|---|---|---|
| description polystyrene, sterile (γ-irradiated), 1 Chamber with Vent Caps, graduations, case of 8 | description clear polystyrene, sterile, graduations, high-density polyethylene cap, case of 2 | description Tissue Culture (TC)-treated surface, clear polystyrene, sterile, graduations, high-density polyethylene cap, case of 5 | description 5 Chamber with Vent Caps, PS, TC-treated, w/ graduations, sterile, 2/cs |
| binding type low binding surface | binding type - | binding type Tissue Culture (TC)-treated surface | binding type Tissue Culture (TC)-treated surface |
| sterility sterile (γ-irradiated) | sterility sterile | sterility sterile | sterility sterile |
| manufacturer/tradename Corning 3303 | manufacturer/tradename Corning 3272 | manufacturer/tradename Corning 3269 | manufacturer/tradename Corning 3319 |
| material cap (vented), rectangular , straight neck , polystyrene , vent cap, rectangular bottom flask | material clear polystyrene , high-density polyethylene cap | material clear polystyrene , high-density polyethylene cap, rectangular bottom flask, short neck | material orange high-density polyethylene cap, rectangular bottom flask, short neck |
| feature graduations, cap | feature cap vented, graduations | feature cap vented, graduations | feature cap vented, graduations |
| surface area 636 cm2 | surface area 25440 cm2 | surface area 1272 cm2 | surface area 3180 cm2 |
Still not finding the right product?
Explore all of our products under Corning® CellSTACK® cell culture chambers
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?