Wybierz wielkość
| Do Państwa/SKU | Dostępność | Cena netto |
|---|---|---|
50 ea | Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności | 1730,00 zł |
Informacje o tej pozycji
1730,00 zł
material
clear plate, flat bottom clear , flat bottom wells , polystyrene , polystyrene plate, round wells
sterility
sterile
feature
lid, skirt, plate format: 96 well standard
packaging
pack of 5, case of 50
manufacturer/tradename
Corning 3300
size
96 wells
surface area
0.32 cm2 , cell growth area
cell growth area
cell growth area
well volume
360 μL
well working volume
75-200 μL
suitability
suitable for (cell cuture)
binding type
CellBIND® surface
General description
- Flat bottoms with 360 μL total volume
- Recommended working volumes of 75 to 200 μL
- CellBIND® surface treated for optimal cell attachment
- Nonreversable lids with condensation rings to reduce contamination
- Sterilized by gamma radiation and certified nonpyrogenic
- Individual alphanumeric codes for well identification
Legal Information
1 of 1
Ta pozycja | |||
|---|---|---|---|
| size 96 wells | size 96 wells | size 96 wells | size 96 wells |
| suitability suitable for (cell cuture) | suitability suitable for (cell cuture) | suitability suitable for (cell cuture) | suitability suitable for (cell culture) |
| material clear plate, flat bottom wells , polystyrene plate, flat bottom clear , round wells | material black plate, black polystyrene , flat bottom clear , flat bottom wells , polystyrene plate, round wells | material flat bottom clear wells, flat bottom wells , polystyrene plate, white polystyrene plate | material flat bottom , white bottom, white polystyrene |
| packaging pack of 5, case of 50 | packaging pack of 5, case of 50 | packaging case of 100 plates, pack of 5, case of 50 | packaging bag of 20 |
| well working volume 75-200 μL | well working volume 75-200 μL | well working volume 75-200 μL | well working volume - |
| surface area 0.32 cm2 , cell growth area | surface area 0.32 cm2 , cell growth area | surface area 0.32 cm2 , cell growth area | surface area - |
Still not finding the right product?
Explore all of our products under Corning® 96 Well CellBIND® Microplates
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?