Zmień widok
| Do Państwa/SKU | Dostępność | Cena netto |
|---|---|---|
2 ea | Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności | 4170,00 zł 3544,50 zł |
Informacje o tej pozycji
NACRES:
NA.75
UNSPSC Code:
41105108
3544,50 zł
Cena katalogowa4170,00 złZaoszczędź 15%Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności
Pomoc techniczna
Pozwól nam pomócdescription
18 in. C-Flex® tubing
sterility
sterile
packaging
pkg of 2 ea
manufacturer/tradename
Corning 10043
General description
Corning® HYPERStack disposable tubing set is sterile C-Flex® tubing compatible for use with 850 cm2 polystyrene roller bottle. The tubing features PureFit tube clamp system, 0.2 µm filter and MPC connector with cap.
Features and Benefits
- It has flexible tubing with a heat seal and chemical resistant
- Easy Grip, threaded cap with male medical grade polycarbonate connector
- Autoclavable with PureFit tube clamp system
- Material is free from animal origin with a polytetrafluoroethylene (PTFE) polymer filter vent
- Sterilized, with a Sterility Assurance Level (SAL) of 10-6
Legal Information
C-Flex is a registered trademark of Saint-Gobain Performance Plastics Corp.
Corning is a registered trademark of Corning, Inc.
HYPERStack is a registered trademark of Corning, Inc.
1 of 1
Ta pozycja | |||
|---|---|---|---|
| description with 850 cm2 polystyrene roller bottle, sterile | description for use with glass bottle, sterile | description 3/8”ID Thermoelastic Tubing, 4 Aseptic Connectors, sterile | description - |
| packaging pkg of 2 ea | packaging pkg of 2 ea | packaging pkg of 2 ea | packaging pkg of 2 ea |
| manufacturer/tradename Corning 10043 | manufacturer/tradename Corning 10042 | manufacturer/tradename Corning 3235 | manufacturer/tradename - |
| sterility sterile | sterility sterile | sterility sterile | sterility - |
| description 18 in. C-Flex® tubing | description 18 in. C-Flex® tubing | description - | description - |
Still not finding the right product?
Explore all of our products under Corning® HYPERStack® disposable tubing set
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?