Przejdź do
B95a
NOTE: Both the cell line and DNA from the cell line may beavailable for this product. Please choose -1VL or VIAL for cells, or -DNA-5UGfor DNA, 1092505, cotton-top tamarin monkey blood, Not specified
Wybierz wielkość
Zmień widok
Rozmiar/SKUDostępnośćCena netto
Informacje o tej pozycji
UNSPSC Code:
41106514
Biological source:
cotton-top tamarin monkey blood
Growth mode:
Semi-adherent
Karyotype:
Not specified
Morphology:
Not specified
Manufacturer/tradename:
ECACC
Informacje o cenach i dostępności nie są obecnie dostępne.
Pomoc techniczna
Pozwól nam pomóc
biological source
cotton-top tamarin monkey blood
description
Convention on the International Trade in Endangered Species of Wild Fauna and Flora (CITES)
manufacturer/tradename
ECACC
packaging
tube of 5 μg 01092505-DNA-5UG, pkg of vial of cells 01092505-1VL
growth mode
Semi-adherent
karyotype
Not specified
morphology
Not specified
receptors
Not specified
Biochem/physiol Actions
B-lymphoblastoid, EBV-transformed
B95a is a derivative of the cell line B95-8 and was adapted to growth in adherence to the substrate surface. The parent line B95-8 was derived from a cotton-top tamarin (Saguinas oedipus). The B-lymphoblastoid cells are transformed with Epstein-Barr virus (EBV). B95a has been reported to be 10,000-fold more sensitive to measles virus present in clinical specimens than Vero cells and are useful host cells for the isolation of measles virus.
Packaging
NOTE: Both the cell line and DNA from the cell line may beavailable for this product. Please choose -1VL or VIAL for cells, or -DNA-5UGfor DNA.
Preparation Note
DMEM (D6546) + 2 mM L-Glutamine (G7513) + 5% FBS / FCS (F2442) (for growth medium) or 2% FBS / FCS (F2442) (for maintenance medium).
Split sub-confluent cultures (70-80%) 1:3 every 3-4 days using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37 °C; cells grow initially as a monolayer, but will pile up and detach in maintenance medium.
Other Notes
Cultures from HPA Culture Collections and supplied by Sigma are for research purposes only. Enquiries regarding the commercial use of a cell line are referred to the depositor of the cell line. Some cell lines have additional special release conditions such as the requirement for a material transfer agreement to be completed by the potential recipient prior to the supply of the cell line. Please view the Terms & Conditions of Supply for more information.
This cell line is part of the European Collection of Authenticated Cell Cultures (ECACC), an international repository managed by the United Kingdom Health Security Agency (UKHSA). No licensing agreement is required when either this cell line or the DNA extracted from it are used for internal research purposes only. Any other use of these products is prohibited without the express written permission of UKHSA. Inquiries regarding authorized use of this cell line or its genomic DNA may be directed to culturecollections@ukhsa.gov.uk.
Porównaj podobne pozycje
Wyświetl pełne porównanie
Pokaż różnice
1 of 1
Ta pozycja
CB_85102918CB_85020205CB_85061107
vibrant-m
B95a
vibrant-m
MA104
vibrant-m
Vero76
vibrant-m
T24/83
description
NOTE: Both the cell line and DNA from the cell line may beavailable for this product. Please choose -1VL or VIAL for cells, or -DNA-5UGfor DNA, 1092505, cotton-top tamarin monkey blood, Not specified
description
NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA, 85102918, African green monkey kidney, Epithelial
description
NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA, 85020205, African green monkey kidney, Epithelial
description
NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA, 85061107, human bladder, Epithelial
description
Convention on the International Trade in Endangered Species of Wild Fauna and Flora (CITES)
description
Convention on the International Trade in Endangered Species of Wild Fauna and Flora (CITES)
description
Convention on the International Trade in Endangered Species of Wild Fauna and Flora (CITES)
description
-
biological source
cotton-top tamarin monkey blood
biological source
African green monkey kidney
biological source
African green monkey kidney
biological source
human bladder
growth mode
Semi-adherent
growth mode
Adherent
growth mode
Adherent
growth mode
Adherent
morphology
Not specified
morphology
Epithelial
morphology
Epithelial
morphology
Epithelial
manufacturer/tradename
ECACC
manufacturer/tradename
ECACC
manufacturer/tradename
ECACC
manufacturer/tradename
ECACC
packaging
tube of 5 μg 01092505-DNA-5UG, pkg of vial of cells 01092505-1VL
packaging
tube of 5 μg 85102918-DNA-5UG, pkg of vial of cells 85102918-1VL
packaging
tube of 5 μg 85020205-DNA-5UG, pkg of vial of cells 85020205-1VL
packaging
tube of 5 μg 85061107-DNA-5UG, pkg of vial of cells 85061107-1VL
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)