Merck
C3858
Scienceware® cloning cylinders, polystyrene
I.D. × H 9.5 mm × 11 mm
Synonim(y):
cloning cylinder, cloning equipment, 378470300, F37847-0300, clone isolation chamber, cloning flask
Informacje o tej pozycji
NACRES:
NB.22
UNSPSC Code:
41106618
Pomoc techniczna
Pozwól nam pomóc
sterility
sterile
packaging
pack of 50 ea
manufacturer/tradename
Bel-Art F37847-0300
I.D. × H
9.5 mm × 11 mm
General description
To isolate a clone, sterile grease (not included) is applied to bottom edge of cylinder and inverted over clone of choice. Provides an isolation chamber to trypsinize and recover clonal colonies.
Legal Information
Scienceware is a registered trademark of Bel-Art Products, a division of SP
Still not finding the right product?
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)