Informacje o tej pozycji
NACRES:
NB.22
UNSPSC Code:
41122107
Material:
PET membrane
Pore size:
0.4 μm
Sterility:
sterile
Binding type:
non-treated surface
material
PET membrane
description
cellGrade(TM) plus
sterility
sterile
packaging
pack of 12 ea (strips of 4 inserts, induvidullay packed)
manufacturer/tradename
BRAND 782911
size
6 well
pore size
0.4 μm
binding type
non-treated surface
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
BRANDplates is a registered trademark of BRAND GMBH + CO KG
Still not finding the right product?
Explore all of our products under BRAND® Insert strips 9mm for 24 x 6 well BRANDplates®
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?