Informacje o tej pozycji
UNSPSC Code:
41103911
Compatibility:
for use with Rotor F-35-6-30 (large rotor bore)
Manufacturer/tradename:
Eppendorf® 5427733000 (EU)
Pomoc techniczna
Pozwól nam pomócfeature
holds 1 x 2.6 - 7 mL round-bottom tube and blood collection tubes, large rotor bore
packaging
pack of 2 ea
manufacturer/tradename
Eppendorf® 5427733000 (EU)
technique(s)
centrifugation: suitable
compatibility
for use with Rotor F-35-6-30 (large rotor bore)
General description
Adapters to fit various size tubes and bottles for use with Eppendorf rotor F-35-6-30.
Application
Centrifuge tube adapters serve to enhance the versatility, safety, and cost-effectiveness of existing centrifuge equipment by allowing laboratories to use a wider variety of sample tube sizes with the same rotor and centrifuge model.
Features and Benefits
- Enhanced Versatility: The primary benefit is expanding the application flexibility of a single centrifuge, accommodating diverse sample volumes and types without needing to purchase multiple rotors or centrifuge models.
- Cost-Effective: Using adapters is more economical than investing in a new rotor or centrifuge for every different tube size required.
- Improved Safety and Stability: They ensure tubes are properly balanced and securely held during operation, which minimizes the risk of tube breakage, sample loss, and damage to the centrifuge equipment from imbalance.
- Streamlined Workflows: Adapters allow for seamless integration of different sample preparation protocols, reducing downtime associated with changing out large rotors and increasing overall lab efficiency.
- Maintained Sample Integrity: By ensuring a secure and proper fit, adapters help maintain consistent separation results and sample integrity.
- Maximized G-Force: When used correctly, adapters help ensure that the required relative centrifugal force (RCF or g-force) is applied correctly to the sample for optimal separation.
- Ease of Use and Maintenance: They are typically easy to insert and remove, simplifying the setup process and facilitating easier cleaning of the rotor chamber.
Legal Information
Eppendorf is a registered trademark of Eppendorf SE
Still not finding the right product?
Explore all of our products under Adapter for Eppendorf® F-35-6-30 Rotor
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?