Przejdź do zawartości
3654P
BIOEAZE Tubing and Components
Informacje o tej pozycji
UNSPSC Code:
41103801
Pomoc techniczna
Pozwól nam pomóc
description
Colder Quick Connect
sterility
sterile; γ-irradiated
packaging
pkg of 10 pkg
General description
Female Quick Cap Polycarbonate for 1/4" and 3/8" Male Quick Connect
Packaging
Individually Bagged
Legal Information
BIOEAZE is a trademark of Sigma-Aldrich Co. LLC
Still not finding the right product?
Explore all of our products under BIOEAZE Tubing and Components
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)