Wybierz wielkość
| Rozmiar/SKU | Dostępność | Cena netto |
|---|---|---|
1 ea | Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności | 778,00 zł |
Informacje o tej pozycji
778,00 zł
material
glass (Clear), round bottom flask, three angled neck
joint
joint: ST/NS24/40
sterility
non-sterile
feature
closure type Standard Taper Joint
packaging
case of 1
manufacturer/tradename
Sigma-Aldrich
capacity
250 mL
Legal Information
1 of 1
Ta pozycja | |||
|---|---|---|---|
| description angled, capacity 250 mL, joint: ST/NS 24/40 | description angled, capacity 500 mL, joint: ST/NS 24/40 | description vertical, capacity 250 mL, joint: ST/NS 24/40 | description angled, capacity 1,000 mL, joint: ST/NS 24/40 |
| capacity 250 mL | capacity 500 mL | capacity 250 mL | capacity 1,000 mL |
| manufacturer/tradename Sigma-Aldrich | manufacturer/tradename Sigma-Aldrich | manufacturer/tradename Sigma-Aldrich | manufacturer/tradename Sigma-Aldrich |
| joint joint: ST/NS24/40 | joint joint: ST/NS24/40 | joint joint: ST/NS24/40 | joint joint: ST/NS24/40 |
| material three angled neck , glass (Clear), round bottom flask | material three angled neck , round bottom flask, glass (Clear) | material three-neck , three-neck (Vertical), round bottom flask, borosilicate glass | material three angled neck , glass (Clear), round bottom flask |
| packaging case of 1 | packaging case of 1 | packaging pack of 1 | packaging case of 1 |
| feature closure type Standard Taper Joint | feature closure type Standard Taper Joint | feature closure type Standard Taper Joint | feature closure type Standard Taper Joint |
Still not finding the right product?
Explore all of our products under Aldrich® three-neck round-bottom flask
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?