Przejdź do
94791
SnapIT Ampule Opener
Regular, for 1-2ml, 5-10ml, 10-15ml ampoules
Informacje o tej pozycji
UNSPSC Code:
12352200
General description
SnapITs are easy-to-use, multi-use ampule openers that avoid the possibility of injury sometimes seen when ampoules are opened by hand.
1. INSERT the ampoule lid into your multi-use SnapIT
2. SNAP open the ampoule
3. EJECT the ampoule lid directly into a sharps container
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)