Zaloguj się, aby wyświetlić ceny organizacyjne i kontraktowe.
Informacje o tej pozycji
UNSPSC Code:
12352200
Nazwa produktu
MILLIPLEX Non-Human Primate Cytokine/Chemokine/Growth Factor Panel A SF, tylko Chiny, Configurable Serum Matrix-Free Non-Human Primate Cytokine/Chemokine/Growth Factor 48-Plex Panel
Quality Segment
product line
Milliplex®
species reactivity
nonhuman primates
packaging
pkg of 1 ea
technique(s)
multiplexing: suitable
input
cell culture supernatant(s)
detection method
fluorometric (Luminex® xMAP® technology)
shipped in
wet ice
storage temp.
2-8°C
General description
Cytokines are small proteins secreted by immune cells that regulate immune responses, hematopoiesis, and lymphocyte development, including interleukins, interferons, and chemokines. Growth factors, which promote cell growth and proliferation, share structural and functional similarities with cytokines and act through specific receptors on target cells. Unlike hormones produced by specialized glands, both cytokines and growth factors can be secreted by various cell types, and their tightly regulated expression is crucial; dysregulation can lead to diseases such as infections, autoimmune disorders, and cancer. Research on these molecules is essential for understanding the immune system and developing therapeutic strategies, with non-human primates serving as important models due to their genetic similarity to humans.
Application
Simultaneously analyze up to 48 analytes in non-human primate cell culture supernatant samples.
Analytes included: BCA-1, sCD137, CD40L, Eotaxin, sFASL, FGF-2, Fractalkine, G-CSF, GM-CSF, Granzyme A, Granzyme B, IFNα2, IFNγ, IL-1α, IL-1β, IL-1RA, IL-2, IL-4, IL-5, IL-6, IL-7, IL-8, IL-10, IL-12 (p70), IL-15, IL-16, IL-17A, IL-17E, IL-18, IL-21, IL-22, IL-23, IL-28A, IL-31, IL-33, IP-10, I-TAC, MCP-1, MIG, MIP-1α, MIP-1β, MIP-3α, Perforin, RANTES, TGFα, TNFα, TNFβ, VEGF-A
Analytes included: BCA-1, sCD137, CD40L, Eotaxin, sFASL, FGF-2, Fractalkine, G-CSF, GM-CSF, Granzyme A, Granzyme B, IFNα2, IFNγ, IL-1α, IL-1β, IL-1RA, IL-2, IL-4, IL-5, IL-6, IL-7, IL-8, IL-10, IL-12 (p70), IL-15, IL-16, IL-17A, IL-17E, IL-18, IL-21, IL-22, IL-23, IL-28A, IL-31, IL-33, IP-10, I-TAC, MCP-1, MIG, MIP-1α, MIP-1β, MIP-3α, Perforin, RANTES, TGFα, TNFα, TNFβ, VEGF-A
Features and Benefits
Designed for Non-Human Primate Cell Culture Supernatants: Optimized for analyzing cytokines, chemokines, and growth factors in cell culture supernatants derived from non-human primate models.
Multiplex Capability: Detect up to 48 analytes simultaneously, maximizing data output from minimal sample input.
Compatible with Luminex® Platforms: Seamlessly integrates with Luminex® xMAP® systems.
Supports Translational Research: Enables detailed immune profiling in primate cell models, bridging preclinical studies with human applications.
Multiplex Capability: Detect up to 48 analytes simultaneously, maximizing data output from minimal sample input.
Compatible with Luminex® Platforms: Seamlessly integrates with Luminex® xMAP® systems.
Supports Translational Research: Enables detailed immune profiling in primate cell models, bridging preclinical studies with human applications.
Legal Information
Luminex is a registered trademark of Luminex Corporation
MILLIPLEX is a registered trademark of Merck KGaA, Darmstadt, Germany
xMAP is a registered trademark of Luminex Corp
Disclaimer
For research use only. Not for use in diagnostic procedures.
Label License/Sticker for Assay Product:
By opening the packaging containing this Assay Product (which contains fluorescently labeled microsphere beads authorized by Luminex Corporation) or using this Assay Product in any manner, you are consenting and agreeing to be bound by the End User Terms and Conditions and the End User License Agreement available at http://support.diasorin.com/end-user-terms-and-conditions/. If you do not agree to all of the terms and conditions, you must promptly return this Assay Product for a full refund prior to using it in any manner.
Label License/Sticker for Assay Product:
By opening the packaging containing this Assay Product (which contains fluorescently labeled microsphere beads authorized by Luminex Corporation) or using this Assay Product in any manner, you are consenting and agreeing to be bound by the End User Terms and Conditions and the End User License Agreement available at http://support.diasorin.com/end-user-terms-and-conditions/. If you do not agree to all of the terms and conditions, you must promptly return this Assay Product for a full refund prior to using it in any manner.
Ta strona może zawierać tekst przetłumaczony maszynowo.
Still not finding the right product?
Explore all of our products under MILLIPLEX Non-Human Primate Cytokine/Chemokine/Growth Factor Panel A SF, tylko Chiny
signalword
Danger
Hazard Classifications
Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Skin Sens. 1 - STOT RE 2
target_organs
Respiratory Tract
Klasa składowania
10 - Combustible liquids
wgk
WGK 3
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?
Dokumenty związane z niedawno zakupionymi produktami zostały zamieszczone w Bibliotece dokumentów.



