Przejdź do zawartości
MAVFA4VP2
MAS-100 VF® Portable Air Sampler Validation Protocol
for use with MAS-100 VF® Microbial Air Samplers
Informacje o tej pozycji
NACRES:
JA.11
UNSPSC Code:
41116500
Pomoc techniczna
Pozwól nam pomóc
availability
not available in North America
compatibility
for use with MAS-100 VF® Microbial Air Samplers
General description
MAS-100 VF® Validation Protocol(A4 Format) is designed as a ready to use step by step guide to perform IQ and OQ activities of MAS-100 VF® microbial air sampling systems. The protocol contains an exhaustive list of tests to cover a batch of units to be validated, the test records protocol MAVFA4TR1 including test descriptions and test records must be ordered separately. The protocol follows GMP, EP, USP and JP regulatory guidelines. Not available in North America.
Application
MAS-100 VF® Validation Protocol is used to validate microbial air sampling systems with ease and consistent performance while following regulatory guidelines.
Legal Information
MAS-100 VF is a registered trademark of MBV AG
Still not finding the right product?
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)