Wybierz wielkość
| Do Państwa/SKU | Dostępność | Cena netto |
|---|---|---|
1 ea | Przewidywana wysyłka DZISIAJzKuehne + Nagel Sp. z o.o. | 491,00 zł |
Informacje o tej pozycji
feature
electronic, repair kit
manufacturer/tradename
EZ-Stream®
application(s)
bioburden testing
food and beverages
industrial qc
pharmaceutical
water monitoring
compatibility
for use with EZ-Stream® Vacuum Pump
General description
This spare part shall be replaced when the switch is unable to operate the pump. The design of the system allows fast and easy replacement of this component.
The EZ-Stream® Vacuum Pump is ideal to be used both on work bench and laminar flow hood, where its small footprint saves laboratory space.
Application
- 1 Circuit Board with LED and Tactile Switch
- 1 Connection wire.
- Applicable Screw Set and spacers.
Legal Information
1 of 1
Ta pozycja | |||
|---|---|---|---|
| compatibility for use with EZ-Stream® Vacuum Pump | compatibility for use with EZ-Stream® Vacuum Pump | compatibility for use with EZ-Stream® Vacuum Pump | compatibility for use with EZ-Stream® Vacuum Pump. |
| feature electronic, repair kit | feature repair kit | feature repair kit | feature repair kit |
| application(s) bioburden testing | application(s) bioburden testing | application(s) bioburden testing | application(s) bioburden testing |
| manufacturer/tradename EZ-Stream® | manufacturer/tradename EZ-Stream® | manufacturer/tradename EZ-Stream® | manufacturer/tradename EZ-Stream® |