Merck
71513-M
Floating Rack, Midi
Informacje o tej pozycji
NACRES:
NB.24
UNSPSC Code:
41104900
Manufacturer/tradename:
Novagen®
Pomoc techniczna
Pozwól nam pomóc
manufacturer/tradename
Novagen®
storage condition
OK to freeze
technique(s)
dialysis: suitable
storage temp.
10-30°C
General description
The Floating Rack is a laboratory tool specifically designed to hold D-Tube Dialyzers in an exchange buffer during dialysis. The Mini Floating Rack can accommodate up to nine D-Tube Dialyzer Mini devices simultaneously. To use, insert the dialyzers into the floating rack and place the assembly into the exchange buffer to carry out small-scale dialysis.
Application
The Millipore Floating Rack, Midi may be used to:
  • suspend D-Tube dialyzer containing dialysis buffer, enabling efficient removal of salts
  • keep dialysis tubes submerged in dialysis buffer, ensuring effective desalting of small-volume biological samples prior to downstream applications such as Liquid Chromatography with Tandem Mass Spectrometry (LC-MS/MS) proteomics and inhibition assays.
Legal Information
NOVAGEN is a registered trademark of Merck KGaA, Darmstadt, Germany
Disclaimer
Toxicity: Standard Handling (A)
Klasa składowania
10-13 - German Storage Class 10 to 13
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)