Informacje o tej pozycji
NACRES:
NA.28
UNSPSC Code:
12352202
Form:
lyophilized
Pomoc techniczna
Pozwól nam pomócQuality Segment
form
lyophilized
storage condition
OK to freeze
solubility
soluble (Reconstitute to a concentration >=10 µg/ml with sterile PBS containing >=0.1% HSA or BSA.)
storage temp.
−70°C
General description
Recombinant, human activin A (disulfide-linked homodimer of two 116-amino acid residue βA subunits) expressed in CHO cells. Mature βA-monomer lacks a potential N-linked glycosylation site and has a calculated molecular mass of 13 kDa. Activins are members of the TGF-β superfamily and have been shown to play an important role in the regulation of homonogenesis, cellular homeostasis, and differentiation programs.
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?