Zmień widok
| Do Państwa/SKU | Dostępność | Cena netto |
|---|
Informacje o tej pozycji
UNSPSC Code:
41100000
Informacje o cenach i dostępności nie są obecnie dostępne.
Pomoc techniczna
Pozwól nam pomócAC/DC input
230 V
material
glass
feature
CE compliant
manufacturer/tradename
Heidolph
parameter
20-180 °C temperature
General description
Safety features
- For your protection against splashing water and steam, the operating panel is located in front of the evaporator for safe and easy access.
- Drip shield over display protects user and control panel from spills
- Individual power switches for control panel and heating bath offers additional safety
- Heating bath features non-slip safety handles for safe and easy transport of bath
- Heating bath is constructed of an insulated double wall for you protection against burns and scalding
- Adjustable sliding safety guard hood always aligns with the bath and protects you in case of implosion
- Long life graphite filled PTFE vacuum seal
- Non-sticking vapor tube removes easily eliminating broken glass
- Unique large bore vapor tube offers fast distribution rates
- One piece vapor tube without additional sealing
- Flask ejector presses off sticking evaporation flasks
- Adjust and fix the height of your evaporating flask easily
- For your convenience, the angle of the evaporation flask can be adjusted from 10 ° to 85 ° quickly
- Superior results due to high torque (140 Ncm) drive
- Choice of manual or motor lift
Still not finding the right product?
Explore all of our products under Heidolph Rotary Evaporator, Laborota 4003
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?