Wybierz wielkość
| Rozmiar/SKU | Dostępność | Cena netto |
|---|---|---|
1 ea | Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności | 562,00 zł |
Informacje o tej pozycji
562,00 zł
material
borosilicate glass flask, round bottom , single-neck
joint
joint: ST/NS14/20
packaging
pkg of 1 ea
manufacturer/tradename
Synthware F461050
capacity
50 mL
valve size
0-4 mm
General description
Legal Information
1 of 1
Ta pozycja | |||
|---|---|---|---|
| description capacity 50 mL, joint: ST/NS 14/20, valve size 0-4 mm | description capacity 10 mL, joint: ST/NS 14/20, valve size 0-4 mm | description capacity 500 mL, joint: ST/NS 14/20, valve size 0-4 mm | description capacity 100 mL, joint: ST/NS 14/20, valve size 0-4 mm |
| manufacturer/tradename Synthware F461050 | manufacturer/tradename Synthware F461010 | manufacturer/tradename Synthware F461500 | manufacturer/tradename Synthware F461100 |
| capacity 50 mL | capacity 10 mL | capacity 500 mL | capacity 100 mL |
| material single-neck , borosilicate glass flask, round bottom | material single-neck , borosilicate glass flask, round bottom | material borosilicate glass flask, single-neck , round bottom | material single-neck , round bottom , borosilicate glass flask |
| joint joint: ST/NS14/20 | joint joint: ST/NS14/20 | joint joint: ST/NS14/20 | joint joint: ST/NS14/20 |
| packaging pkg of 1 ea | packaging pkg of 1 ea | packaging pkg of 1 ea | packaging pkg of 1 ea |
| valve size 0-4 mm | valve size 0-4 mm | valve size 0-4 mm | valve size 0-4 mm |
Still not finding the right product?
Explore all of our products under Synthware™ storage flask with standard taper side arm
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?