Merck
SYNF448500
Synthware™ three neck round bottom flask with vertical side necks
500 mL, center joint: ST/NS 29/42, side joint: ST/NS 29/42
Synonim(y):
Synthware F448500, 3-neck round flask
Informacje o tej pozycji
NACRES:
NB.51
UNSPSC Code:
41121804
eCl@ss:
32040501
Capacity:
500 mL, 500 mL
Material:
borosilicate glass flask, round bottom , three-neck (vertical side necks)
Joint:
center joint: ST/NS29/42, side joint: ST/NS29/42
Packaging:
pkg of 1 ea
Pomoc techniczna
Pozwól nam pomóc
material
borosilicate glass flask, round bottom , three-neck (vertical side necks)
joint
center joint: ST/NS29/42, side joint: ST/NS29/42
packaging
pkg of 1 ea
manufacturer/tradename
Synthware F448500
capacity
500 mL, 500 mL
General description
Round bottom with joints. Heavy wall design. All sizes are hand-blown from tubing to ensure uniform wall thickness.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Still not finding the right product?
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)