Przejdź do
BR36407
BRAND® BLAUBRAND® volumetric flask trapezoidal
capacity 25 mL, polypropylene stopper, joint: ST/NS 10/19
Synonim(y):
BLAUBRAND®, Volumetric flasks
Wybierz wielkość
Zmień widok
Rozmiar/SKUDostępnośćCena netto
Informacje o tej pozycji
NACRES:
NB.43
UNSPSC Code:
41121804
eCl@ss:
32040501
Capacity:
25 mL
Material:
polypropylene stopper, transparent borosilicate glass 3.3 flask
Packaging:
pack of 2 ea
Informacje o cenach i dostępności nie są obecnie dostępne.
Pomoc techniczna
Pozwól nam pomóc
material
polypropylene stopper, transparent borosilicate glass 3.3 flask
joint
joint: ST/NS10/19
feature
accuracy class A
packaging
pack of 2 ea
manufacturer/tradename
BRAND 36407
capacity
25 mL
General description
Borosilicate 3.3. To DIN EN ISO 1042. Calibrated to contain (TC, In).
All BLAUBRAND® volumetric flasks are delivered with batch number and enclosed one batch certificate per packing unit of the manufacturer. On demand also available with individual certificate or DAkkS calibration certificate.
Legal Information
BLAUBRAND is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG
Porównaj podobne pozycje
Wyświetl pełne porównanie
Pokaż różnice
1 of 1
Ta pozycja
BR36406BR36402BR36408
description
capacity 25 mL, polypropylene stopper, joint: ST/NS 10/19
description
capacity 20 mL, polypropylene stopper, joint: ST/NS 10/19
description
capacity 2 mL, polypropylene stopper, joint: ST/NS 7/16
description
capacity 50 mL, polypropylene stopper, joint: ST/NS 12/21
capacity
25 mL
capacity
20 mL
capacity
2 mL
capacity
50 mL
manufacturer/tradename
BRAND 36407
manufacturer/tradename
BRAND 36406
manufacturer/tradename
BRAND 36402
manufacturer/tradename
BRAND 36408
feature
accuracy class A
feature
accuracy class A
feature
accuracy class A
feature
accuracy class A (To contain)
material
polypropylene stopper, transparent borosilicate glass 3.3 flask
material
polypropylene stopper, transparent borosilicate glass 3.3 flask
material
polypropylene stopper, transparent borosilicate glass 3.3 flask
material
transparent borosilicate glass 3.3 flask, polypropylene stopper
joint
joint: ST/NS10/19
joint
joint: ST/NS10/19
joint
joint: ST/NS7/16
joint
joint: ST/NS12/21
packaging
pack of 2 ea
packaging
pack of 2 ea
packaging
pack of 2 ea
packaging
pack of 2 ea
Still not finding the right product?
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)