Merck
BR23723
BRAND® spare burette for automatic burette Dr. Schilling
size 25 mL, amber
Synonim(y):
Accessories, Automatic Burettes, Burettes, Dr. Schilling pattern, SILBERBRAND
Informacje o tej pozycji
NACRES:
NB.43
UNSPSC Code:
41121808
eCl@ss:
32070701
Material:
AR-Glass , amber
Pomoc techniczna
Pozwól nam pomóc
material
AR-Glass , amber
description
SILBERBRAND
packaging
pack of 1 ea
manufacturer/tradename
BRAND 23723
L
440 mm
size
25 mL
General description
Spare burette for automatic burette Schilling, made of Boro 3.3 glass, available in various nominal volumes for precise liquid dispensing.
Application
Liquid handling
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Still not finding the right product?
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)