Przejdź do
BR136335
BRAND® stirring rod
high-density polyethylene (with steel core), L × D 400 mm × 6 mm
Synonim(y):
PTFE, Stirring rods
Wybierz wielkość
Zmień widok
Rozmiar/SKUDostępnośćCena netto
1 ea
Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności
108,00 zł
Informacje o tej pozycji
NACRES:
NB.72
UNSPSC Code:
41121802
Manufacturer/tradename:
BRAND 136335
Material:
high-density polyethylene (with steel core)
108,00 zł
Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności
​
Pomoc techniczna
Pozwól nam pomóc
material
high-density polyethylene (with steel core)
description
with steel core
packaging
pack of 1 ea
manufacturer/tradename
BRAND 136335
L × D
400 mm × 6 mm
General description
PTFE spatula-shaped stirring rods designed for efficient mixing and stirring.
The BRAND® stirring rod is designed to be bendable, allowing for customization based on specific applications. The permanent bend ensures that the rod maintains its shape during use, providing flexibility for stirring in various vessel shapes and sizes. This innovative design enhances the efficiency of mixing and blending processes, making it suitable for a wide range of laboratory tasks.
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Porównaj podobne pozycje
Wyświetl pełne porównanie
Pokaż różnice
1 of 1
Ta pozycja
BR136320BR136315BR136325
description
high-density polyethylene (with steel core), L × D 400 mm × 6 mm
description
high-density polyethylene (with steel core), L × D 250 mm × 6 mm
description
high-density polyethylene (with steel core), L × D 200 mm × 6 mm
description
high-density polyethylene (with steel core), L × D 300 mm × 6 mm
manufacturer/tradename
BRAND 136335
manufacturer/tradename
BRAND 136320
manufacturer/tradename
BRAND 136315
manufacturer/tradename
BRAND 136325
material
high-density polyethylene (with steel core)
material
high-density polyethylene (with steel core)
material
high-density polyethylene (with steel core)
material
high-density polyethylene (with steel core)
packaging
pack of 1 ea
packaging
pack of 1 ea
packaging
pack of 1 ea
packaging
pack of 1 ea
description
with steel core
description
with steel core
description
with steel core
description
with steel core
L × D
400 mm × 6 mm
L × D
250 mm × 6 mm
L × D
200 mm × 6 mm
L × D
300 mm × 6 mm
Still not finding the right product?
Explore all of our products under BRAND® stirring rod
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)