Przejdź do zawartości
BMSBR5000E
Everlast rocker with flat mat
AC/DC input 230 V AC, Schuko plug
Informacje o tej pozycji
UNSPSC Code:
41103811
Pomoc techniczna
Pozwól nam pomóc
manufacturer/tradename
Benchmark Scientific BR5000-E
W × D
16 in. × 12 in.
General description
  • Stainless-steel platform
  • Dual platform accessory doubles work surface
  • Cold room and incubator safe
The rocking speed and tilt angle can easily be adjusted
to vary between gentle and aggressive agitation. Both
the standard platform and the (optional) stacking
platform are constructed of stainless steel, preventing
against any rusting or corrosion. The Everlast 247 is
designed to provide years of uninterrupted service.
Legal Information
Everlast is a trademark of Benchmark Scientific, Inc.
Still not finding the right product?
Explore all of our products under Everlast rocker with flat mat
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)