Merck
  • Scanning fluorescent microscopy analysis is applicable for absolute and relative cell frequency determinations.
Scanning fluorescent microscopy analysis is applicable for absolute and relative cell frequency determinations.
Cytometry. Part A : the journal of the International Society for Analytical Cytology (2004-09-08)
József Bocsi, Viktor Sebestyén Varga, Béla Molnár, Ferenc Sipos, Zsolt Tulassay, Attila Tárnok
ABSTRACT
Flow cytometry (FCM) and laser scanning cytometry (LSC) are the routine techniques for fluorescent cell analysis. Recently, we developed a scanning fluorescent microscopy (SFM) technique. This study compares SFM to LSC (two slide-based cytometry, SBC, techniques) and FCM, in experimental and clinical settings. For the relative cell-frequency determinations, HT29 colorectal cancer cells and Ficoll separated blood mononuclear cells (FSBMCs) were serially diluted (from 1:1 to 1:1,000) and measured by each of the three techniques. For the absolute cell number determinations (only for SBC) FSBMCs were smeared on slides, then HT29 cells were placed on the slide with a micromanipulator (5-50 cells). Tumor cells circulating in the peripheral blood were isolated by magnetic separation from clinical blood samples of colorectal cancer patients. All samples were double-stained by CD45 ECD and CAM5.2 FITC antibodies. For slides, TOTO-3 and Hoechst 33258 DNA dyes were applied as nuclear counter staining. In the relative cell frequency determinations, the correlations between the calculated value and measured values by SFM, LSC, and FCM were r(2) = 0.79, 0.62, and 0.84, respectively (for all P < 0.01). In the absolute cell frequency determinations, SFM and LSC correlated to a high degree (r(2) = 0.97; P < 0.01). SFM proved to be a reliable alternative method, providing results comparable to LSC and FCM. SBC proved to be more suitable for rare-cell detection than FCM. SFM with digital slides may prove an acceptable adaptation of conventional fluorescent microscopes in order to perform rare-cell detection.
Sigma-Aldrich
Trypsin-EDTA solution, 0.25%, sterile-filtered, BioReagent, suitable for cell culture, 2.5 g porcine trypsin and 0.2 g EDTA, 4Na per liter of Hanks′ Balanced Salt Solution with phenol red
Pricing and availability is not currently available.