Merck
  • The structure of the covalent adduct formed by the interaction of 3-dimethylamino-1-propyne and the flavine of mitochondrial amine oxidase.
The structure of the covalent adduct formed by the interaction of 3-dimethylamino-1-propyne and the flavine of mitochondrial amine oxidase.
Biochemistry (1976-01-13)
A L Maycock, R H Abeles, J I Salach, T P Singer
PMID2278
ABSTRACT
3-Dimethylamino-1-propyne irreversibly inactivates mitochondrial monoamine oxidase from bovine liver. The inactivation results in the loss of absorption in the 450-500-nm region of the flavine spectrum and a concomitant increase in absorbance at 410 nm. For the enzyme-bound adduct epsilon410 = 28000. The spectral properties of the adduct of the liver enzyme with 3-dimethylamino-1-propyne are similar to those observed when the pig kidney enzyme is inactivated with pargyline (Chuang et al. (1974), J. Biol. Chem. 249, 2381). From a proteolytic digest of the enzyme inactivated with labeled inhibitor a flavine peptide has been isolated which contains 1 mol of inactivator/mol of flavine. The chemical and spectral properties of the adduct are those of compounds containing the structure --N--CH==CH--CH==N+ less than. It was concluded that the flavine-inhibtor adduct is a N-5 substituted dihydroflavine and its structure has been determined.
Sigma-Aldrich
3-Dimethylamino-1-propyne, 97%
Pricing and availability is not currently available.